Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

B-018

Supelco

γ-Butyrolactone solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

format

single component solution

storage temp.

−20°C

SMILES string

O=C1CCCO1

InChI

1S/C4H6O2/c5-4-2-1-3-6-4/h1-3H2

InChI key

YEJRWHAVMIAJKC-UHFFFAOYSA-N

General description

A Certified Snap-N-Spike® Solution appropriate for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology and forensic analysis to urine drug testing. γ-Butyrolactone is a common chemical solvent used in such applications as glue remover and paint stripper. γ-Butyrolactone is commonly abused recreationally for its euphoric and sedative effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Westendorf et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(10), 3853-3858 (2013-02-23)
The rapid reorganization of the actin cytoskeleton in response to external stimuli is an essential property of many motile eukaryotic cells. Here, we report evidence that the actin machinery of chemotactic Dictyostelium cells operates close to an oscillatory instability. When
Axel Bidon-Chanal et al.
European journal of medicinal chemistry, 60, 479-489 (2013-01-29)
Glycogen synthase kinase 3β (GSK-3β) is widely recognised as a relevant player in the pathogenesis of several highly prevalent disorders such as Alzheimer's disease, mood disorders, diabetes and cancer. Therefore, this enzyme constitutes a highly attractive therapeutic target for the
David Santos et al.
The Journal of organic chemistry, 78(4), 1519-1524 (2013-01-24)
An enantioselective approach to (-)-isoavenaciolide was achieved starting from 1-undecyn-3-ol. The synthesis relied upon the preparation of a chiral 4-silyloxy-2-alkenylborane by hydroboration of a protected 2,3-allenol and subsequent stereoselective addition to 2-thiophenecarboxaldehyde.
Zheng-Yuan Su et al.
Chemical research in toxicology, 26(3), 477-485 (2013-02-28)
Cancer development has been linked to epigenetic modifications of cancer oncogenes and tumor suppressor genes; in advanced metastatic cancers, severe epigenetic modifications are present. We previously demonstrated that the progression of prostate tumors in TRAMP mice is associated with methylation
R van Vugt et al.
Acta anaesthesiologica Belgica, 63(3), 139-141 (2013-02-13)
We present two cases of GBL intoxication. Patient A is a 45-year old man who lost consciousness after drinking a clear unknown liquid. He was brought to the Emergency Departement (ED) after he was intubated. His partner notified the liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service