Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-914

Supelco

7-Aminonitrazepam solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O
CAS Number:
Molecular Weight:
251.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

O=C1NC2=CC=C(N)C=C2C(C3=CC=CC=C3)=NC1

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)

InChI key

OYOUQHVDCKOOAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Aminonitrazepam is a major urinary metabolite of nitrazepam, a benzodiazepine used to treat insomnia and sold under the trade names Mogadon and Alodorm. This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine drug monitoring of nitrazepam use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B J Hart et al.
Methods and findings in experimental and clinical pharmacology, 10(1), 21-26 (1988-01-01)
The stability of some selected benzodiazepines in saliva has been studied. The benzodiazepines nitrazepam and clonazepam were found to be unstable in saliva at room temperature and nitrazepam was converted into 7-aminonitrazepam. The conversion rate of nitrazepam was strongly dependent
S Takeno et al.
Toxicology and applied pharmacology, 121(2), 233-238 (1993-08-01)
This study was designed to evaluate the developmental toxicity of nitrazepam (NZ) in Sprague-Dawley rats and ICR mice and to determine the metabolic factors which modulate susceptibility to the developmental effects of NZ. Rats were treated orally with a single
Fumio Moriya et al.
Forensic science international, 131(2-3), 108-112 (2003-02-19)
We report a case of nitrazepam poisoning in which the distribution of nitrazepam and 7-aminonitrazepam was determined in body fluids and tissues. A 52-year-old woman was found dead in a shallow ditch (approximately 5 cm in depth), in the winter.
M Tomita et al.
Journal of chromatography, 621(2), 249-255 (1993-11-24)
We applied micellar electrokinetic capillary chromatography to simultaneous separation and determination of nitrazepam and its major metabolites, 7-aminonitrazepam and 7-acetamidonitrazepam, in spiked urine. Prior to electrophoresis, the three compounds were successfully extracted from the spiked urine with commercial disposable solid-phase
J Feely et al.
Irish journal of medical science, 168(1), 8-9 (1999-03-31)
An alarming increase in the misuse/abuse of nitrobenzodiazepine derivatives, especially flunitrazepam, prompted us to establish reliable analytical protocols for their routine detection. Whilst the parent drugs are readily available from a number of commercial sources, it was found difficult to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service