Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-159

Supelco

Asenapine-D3 maleate solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13D3ClNO · C4H4O4
Molecular Weight:
404.86
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC=C2C([C@](CN(C([2H])([2H])[2H])C3)([H])[C@@]3([H])C(C=CC=C4)=C4O2)=C1.O=C(O)/C=C\C(O)=O

General description

This Snap-N-Shoot® internal standard is suitable for many LC/MS and GC/MS applications including forensic analysis, clinical toxicology, pharmaceutical research, or isotope dilution methods. Asenapine is a new atypical antipsychotic developed for the treatment of schizophrenia and acute mania associated with bipolar disorder. The drug is marketed under the trade names Saphris® and Sycrest®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Saphris is a registered trademark of MSD
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service