Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-151

Supelco

Atorvastatin Impurity Mixture solution

certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H47FN2O5
Molecular Weight:
654.81
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Atorvastatin Calcium, 50 μg/mL (each analyte)
Atorvastatin Related Compound A, 50 μg/mL (each analyte)
Atorvastatin Related Compound B, 50 μg/mL (each analyte)
Atorvastatin Related Compound C, 50 μg/mL (each analyte)
Atorvastatin Related Compound I, 50 μg/mL (each analyte)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

multi-component solution

Storage temp.

−20°C

General description

Atorvastatin Calcium is a synthetic cholesterol lowering agent,[2] <https://www.sigmaaldrich.com/catalog/product/sial/phr1422?lang=en&region=US> which acts as an inhibitor of 3-hydroxy-3-methyl glutaryl coenzyme A reductase. It is widely used in combination with amlodipine besylate as a medication in the treatment of vasospastic angina, hypertension, chronic stable angina and primary dysbetalipoproteinemia. This certified solution standard contains multiple impurities, including Related Compounds A, B, C, and I as well as Atorvastatin itself. This standard can be used for multiple HPLC applications including pharmaceutical testing, research, and Monograph testing methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service