Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-134

Supelco

Acetaminophen (Paracetamol) Impurity Mixture solution

25 μg/mL in methanol (each analyte), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

25 μg/mL in methanol (each analyte)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

multi-component solution

storage temp.

−20°C

General description

Acetaminophen is formed as a metabolic product of phenacetin and is a well-known drug used in the treatment of pain and fever due to its effective central analgesic and antipyretic properties. It is also the only analgesic drug that is non-opioid and available in the oral, rectal, and intravenous formulation forms. It has been posited to inhibit the synthesis of prostaglandins and exhibit a mode of action similar to nonsteroidal anti-inflammatory drugs (NSAIDs).[1][2][3][4][5]

Application

Acetaminophen Impurity Mixture solution may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[6]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetaminophen Related Compound B, Pharmaceutical Secondary Standard; Certified Reference Material
    Acetaminophen Related Compound C, Pharmaceutical Secondary Standard; Certified Reference Material
    Acetaminophen Related Compound D, Pharmaceutical Secondary Standard; Certified Reference Material
    Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
    Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Acetaminophen poisoning and toxicity
Rumack BH and Matthew H
Pediatrics, 55(6), 871- 876 (1975)
Acetaminophen-induced hepatotoxicity
James LP, et al.
Drug Metabolism and Disposition, 31(12), 1499- 1506 (2003)
Central analgesic effect of acetaminophen but not of aspirin
Piletta P, et al.
Clinical Pharmacology and Therapeutics, 49(4), 350- 354 (1991)
Paracetamol (Acetaminophen): mechanisms of action
Anderson BJ
Pediatric Anesthesia, 18(10), 915- 921 (2008)
Intravenous paracetamol (acetaminophen)
Duggan ST and Scott LJ
Drugs, 69(1), 101- 113 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service