Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-108

Supelco

5-APB-D5 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8D5NO · HCl
Molecular Weight:
216.72
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

NC([2H])(C([2H])([2H])[2H])C([2H])C1=CC=C2C(C=CO2)=C1.Cl

InChI

1S/C11H13NO.ClH/c1-8(12)6-9-2-3-11-10(7-9)4-5-13-11;/h2-5,7-8H,6,12H2,1H3;1H/i1D3,6D,8D;

InChI key

NFQIXCQPRSHUTF-QJCNSVPISA-N

General description

5-APB, a derivative of MDA, is a designer drug and stimulant sometimes found in certain bath salt blends. This certified Snap-N-Spike® Solution is suitable for use as an internal standard in amphetamine GC/MS or LC/MS testing applications from forensic analysis and clinical toxicology to urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hajnalka Nyitrai et al.
Cell reports, 31(10), 107712-107712 (2020-06-11)
Neurons face unique transport challenges. They need to deliver cargo over long axonal distances and to many presynaptic nerve terminals. Rab GTPases are master regulators of vesicular traffic, but essential presynaptic Rabs have not been identified. Here, we find that
Richard G Held et al.
Neuron, 107(4), 667-683 (2020-07-04)
Presynaptic CaV2 channels are essential for Ca2+-triggered exocytosis. In addition, there are two competing models for their roles in synapse structure. First, Ca2+ channels or Ca2+ entry may control synapse assembly. Second, active zone proteins may scaffold CaV2s to presynaptic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service