Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A-107

Supelco

5-API (5-IT) solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2
CAS Number:
Molecular Weight:
174.24
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C11H14N2/c1-8(12)6-9-2-3-11-10(7-9)4-5-13-11/h2-5,7-8,13H,6,12H2,1H3

InChI key

AULGMISRJWGTBA-UHFFFAOYSA-N

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for 5-API (5-IT) LC/MS or GC/MS testing methods in clinical toxicology, forensic analysis or urine drug testing applications. 5-API (5-IT), also known as 5-(2-Aminopropyl)indole, is an indole derivative with stimulant effects. This designer drug has been found as an adulterant in other illicit drugs including MDMA, and among "balth salt" products labeled as "Benzo Fury".

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service