Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

999998C

Avanti

06:0 Diether PC

Avanti Research - A Croda Brand 999998C

Synonym(s):

1,2-di-O-hexyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H44NO6P
CAS Number:
Molecular Weight:
425.54
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (999998C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999998C

concentration

10 mg/mL (999998C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCC)COCCCCCC)=O

General description

06:0 Diether PC (phosphocholine) is a glycerophospholipid having two hexane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.

Application

06:0 Diether PC or 1,2-di-O-hexyl-sn-glycero-3-phosphocholine might be used:
  • in bicelle media to measure the H—C one-bond residual dipolar couplings for trisaccharides, Lewis X (Lex) and Lewis A (Lea) methyl glycosides using nuclear magnetic resonance (NMR) residual dipolar couplings
  • to compose isotropically tumbling small bicelles, to study its size and morphology by 31P NMR and atomic force microscopy (AFM)
  • in bicelle preparation for NMR studies

Biochem/physiol Actions

06:0 Diether PC (phosphocholine) is useful for bicelles preparation. Bicelles can be integrated into standard crystallization protocols and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation. Incorporating 06:0 Diether PC in bicelles results in bicelles like conventional phospholipid bicelles but with improved chemical stability.

Packaging

5 mL Clear Glass Sealed Ampule (999998C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed N Triba et al.
European biophysics journal : EBJ, 35(3), 268-275 (2005-09-28)
A protocol is described for the reconstitution of a transmembrane beta-barrel protein domain, tOmpA, into lipid bicelles. tOmpA is the largest protein to be reconstituted in bicelles to date. Its insertion does not prevent bicelles from orienting with their plane
Hongwei Wu et al.
Biochimica et biophysica acta, 1798(3), 482-488 (2009-11-17)
Aqueous phospholipid mixtures that form bilayered micelles (bicelles) have gained wide use by molecular biophysicists during the past 20 years for spectroscopic studies of membrane-bound peptides and structural refinement of soluble protein structures. Nonetheless, the utility of bicelle systems may
Hugo F Azurmendi et al.
Biopolymers, 63(2), 89-98 (2002-01-12)
The conformations of the histo-blood group carbohydrate antigens Lewis X (Le(x)) and Lewis A (Le(a)) were studied by NMR measurements of one-bond C-H residual dipolar couplings in partially oriented liquid crystal solutions. A strategy for rapid calculation of the difference
John M Dean et al.
Protein & cell, 9(2), 196-206 (2017-05-20)
Ether lipids, such as plasmalogens, are peroxisome-derived glycerophospholipids in which the hydrocarbon chain at the sn-1 position of the glycerol backbone is attached by an ether bond, as opposed to an ester bond in the more common diacyl phospholipids. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service