Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

999995C

Avanti

Edelfosine

1-O-octadecyl-2-O-methyl-sn-glycero-3-phosphocholine, chloroform

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H58NO6P
CAS Number:
Molecular Weight:
523.73
UNSPSC Code:
12352211
NACRES:
NA.25
Product 999995C is not currently sold in your country. Contact Technical Service

packaging

pkg of 1 × 1 mL (999995C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999995C

concentration

5 mg/mL (999995C-5mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC)COCCCCCCCCCCCCCCCCCC)=O

InChI

1S/C10H24NO5P/c1-6-10(14-5)9-16-17(12,13)15-8-7-11(2,3)4/h10H,6-9H2,1-5H3/t10-/m0/s1

InChI key

GVMCXWJIRSIWFJ-JTQLQIEISA-N

Application

Edelfosine or 1-O-octadecyl-2-O-methyl-sn-glycero-3-phosphocholine might be used:
  • as a non-hydrolysable LysoPC (phospholipid) analog for analyzing its ability to block sexual commitment in Plasmodium falciparum[1]
  • in multilamellar vesicle preparation to study its effect on model membranes[2]
  • in the selection and screening of mutagenized cells, having the ability to inhibit the transport of alkylphosphocholine drugs across the plasma membrane[3]

Biochem/physiol Actions

Edelfosine acts as a precursor for alkyl-lysophospholipids. It possesses apoptotic action against several cancer cells such as prostate, leukemia, brain and lung tumors. It is associated with cellular transport system, signaling transducing systems, cytokine synthesis and lipid metabolism. Edelfosine is known to control intracellular calcium levels. It is not mutagenic[2] and its anti-tumor action requires its incorporation into the cell.[4] Edelfosine is also found to block the replication of human immunodeficiency virus type I (HIV-I).[2]

Packaging

5 mL Clear Glass Sealed Ampule (999995C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessio Ausili et al.
The journal of physical chemistry. B, 112(37), 11643-11654 (2008-08-21)
The effect of edelfosine (1- O-octadecyl-2- O-methyl-rac-glycero-3-phosphocholine or ET-18-OCH3) on model membranes containing 1-palmitoyl-2-oleoyl- sn-glycero-3-phosphocholine/sphingomyelin/cholesterol (POPC/SM/cholesterol) was studied by several physical techniques. The sample POPC/SM (1:1 molar ratio) showed a broad phase transition as seen by DSC, X-ray diffraction, and
Pamela K Hanson et al.
The Journal of biological chemistry, 278(38), 36041-36050 (2003-07-05)
The alkylphosphocholine class of drugs, including edelfosine and miltefosine, has recently shown promise in the treatment of protozoal and fungal diseases, most notably, leishmaniasis. One of the major barriers to successful treatment of these infections is the development of drug
Bruno M Castro et al.
The journal of physical chemistry. B, 117(26), 7929-7940 (2013-06-07)
Edelfosine (1-O-octadecyl-2-O-methyl-sn-glycero-phosphocholine) and miltefosine (hexadecylphosphocholine) are synthetic alkylphospholipids (ALPs) that are reported to selectively accumulate in tumor cell membranes, inducing Fas clustering and activation on lipid rafts, triggering apoptosis. However, the exact mechanism by which these lipids elicit these events
Nicolas M B Brancucci et al.
Cell, 171(7), 1532-1544 (2017-11-14)
Transmission represents a population bottleneck in the Plasmodium life cycle and a key intervention target of ongoing efforts to eradicate malaria. Sexual differentiation is essential for this process, as only sexual parasites, called gametocytes, are infective to the mosquito vector. Gametocyte production
Alessio Ausili et al.
Langmuir : the ACS journal of surfaces and colloids, 34(28), 8333-8346 (2018-06-21)
Edelfosine is an anticancer drug with an asymmetric structure because, being a derivative of glycerol, it possesses two hydrophobic substituents of very different lengths. We showed that edelfosine destabilizes liquid-ordered membranes formed by either 1-palmitoyl-2-oleoyl- sn-glycero-3-phosphocholine, sphingomyelin (SM), and cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service