Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

999994C

Avanti

12:0 Diether PC

Avanti Research - A Croda Brand 999994C

Synonym(s):

1,2-di-0-dodecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H68NO6P
CAS Number:
Molecular Weight:
593.86
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (999994C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999994C

concentration

10 mg/mL (999994C-10mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCC)COCCCCCCCCCCCC)=O

General description

12:0 Diether PC (phosphocholine) is a glycerophospholipid having two dodecane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.[1]

Application

12:0 Diether PC or 1,2-di-O-dodecyl-sn-glycero-3-phosphocholine might be used in small micelles for nuclear magnetic resonance (NMR) experiments to study the interaction of peptide with model membranes.[2]

Biochem/physiol Actions

12:0 Diether PC (phosphocholine) is found to be a simpler membrane model and is highly compatible with nuclear magnetic resonance (NMR).[2] It is useful in bicelles formation. Bicelles can be integrated into standard crystallization protocols and in contrast to micelles, bicelles maintain the protein in a more native bilayer environment allowing proteins to be captured in a more biologically relevant orientation.

Packaging

5 mL Clear Glass Sealed Ampule (999994C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of a peptide derived from C-terminus of human TRPA1 channel with model membranes mimicking the inner leaflet of the plasma membrane
Witschas K, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(5), 1147-1156 (2015)
Structural and functional roles of ether lipids
Dean JM and Lodhi IJ
Protein & cell, 9(2), 196-206 (2018)

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service