Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

999985C

Avanti

4ME 16:0 Diether PE

Avanti Research - A Croda Brand 999985C

Synonym(s):

1,2-di-O-phytanyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H94NO6P
CAS Number:
Molecular Weight:
776.20
UNSPSC Code:
12352211

form

liquid

packaging

pkg of 1 × 1 mL (999985C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999985C

concentration

10 mg/mL (999985C-10mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OCCC(C)CCCC(C)CCCC(C)CCCC(C)C)COCCC(C)CCCC(C)CCCC(C)CCCC(C)C

General description

4ME 16:0 Diether PE or 1,2-di-O-phytanyl-sn-glycero-3-phosphoethanolamine is a class of glycerophospholipids having two phytane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has ethanolamine as the alcohol moiety, attached to the phosphate group.

Application

4ME 16:0 Diether PE or 1,2-di-O-phytanyl-sn-glycero-3-phosphoethanolamine is suitable:
  • for nanoformulation of liposomal docosahexaenoic acid (DHA) and ghost (no DHA, control) liposomes
  • for the preparation of archaeal-like lipid bilayers to examine the effect of humidity and temperature on it
  • as an internal standard in tandem mass spectrometry/mass spectrometry/mass spectrometry (MS3) for the analysis of polyunsaturated aminophospholipid molecular species
  • as an internal standard in lipid extraction using methyl-tert-butylether (MTBE)/methanol protocol

Packaging

5 mL Clear Glass Sealed Ampule (999985C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Celine Fernandez et al.
PloS one, 8(8), e71846-e71846 (2013-08-24)
We tested whether characteristic changes of the plasma lipidome in individuals with comparable total lipids level associate with future cardiovascular disease (CVD) outcome and whether 23 validated gene variants associated with coronary artery disease (CAD) affect CVD associated lipid species.
Analysis of polyunsaturated aminophospholipid molecular species using isotope-tagged derivatives and tandem mass spectrometry/mass spectrometry/mass spectrometry
Berry KAZ and Murphy RC
Analytical Biochemistry, 349(1), 118-128 (2006)
Marta Salvador-Castell et al.
International journal of molecular sciences, 21(5) (2020-03-12)
Archaea, the most extremophilic domain of life, contain ether and branched lipids which provide extraordinary bilayer properties. We determined the structural characteristics of diether archaeal-like phospholipids as functions of hydration and temperature by neutron diffraction. Hydration and temperature are both
Christine G Skibinski et al.
Chemico-biological interactions, 252, 1-8 (2016-04-05)
The risk of breast cancer can be influenced by certain dietary components, such as the amount and type of dietary fatty acids ingested. Docosahexaenoic acid (DHA), a component of fish oil, is known to suppress rat mammary carcinogenesis, reduce cell
Marta Salvador-Castell et al.
Biochimica et biophysica acta. Biomembranes, 1862(2), 183130-183130 (2019-11-18)
It is now well established that cell membranes are much more than a barrier that separate the cytoplasm from the outside world. Regarding membrane's lipids and their self-assembling, the system is highly complex, for example, the cell membrane needs to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service