Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900122P

Avanti

15-keto PGF2α

powder

Synonym(s):

15-keto PGF2α

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H32O5
CAS Number:
Molecular Weight:
352.47
UNSPSC Code:
12352211

assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of (pkg of 1 X 1 mg (900122P-1MG))

lipid type

neutral lipids
neutral glycerides

shipped in

dry ice

Storage temp.

−20°C

SMILES string

O[C@@H]1C[C@H](O)[C@H](C/C=C\CCCC(O)=O)[C@H]1/C=C/C(CCCCC)=O

Packaging

5 mL Amber Glass Screw Cap Vial (900122P-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service