Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900116P

Avanti

PGF1ß-d9

powder

Synonym(s):

Prostaglandin F1β-d9

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H27D9O5
CAS Number:
Molecular Weight:
365.55
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

pkg of (pkg of 1 X 100 ug (900116P-100UG))
pkg of (pkg of 1 X 50 ug (900116P-50ug))
pkg of (pkg of 1 X 500 ug (900116P-500UG))

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@@H](O)CC([2H])(C([2H])(C([2H])(C([2H])([2H])[2H])[2H])[2H])[2H])[C@H](O)C1

InChI

1S/C20H36O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-19,21-23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,18+,19+/m0/s1/i1D3,2D2,3D2,6D2

InChI key

DZUXGQBLFALXCR-NCLJNVOJSA-N

Packaging

2 mL Amber Glass Sealed Ampule (900116P-50UG)
2 mL Amber Glass Sealed Ampule (900116P-100UG)
2 mL Amber Glass Sealed Ampule (900116P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service