900115P
Avanti
PGA1
Prostaglandin A1, powder
Synonym(s):
7-[(1R,2S)-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopent-3-en-1-yl]heptanoic acid
Select a Size
Select a Size
About This Item
Recommended Products
form
powder
packaging
pkg of 1 × 1 mg (900115P-1mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 900115P
shipped in
dry ice
storage temp.
−20°C
SMILES string
O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)C=C1
InChI
1S/C20H32O4/c1-2-3-6-9-17(21)14-12-16-13-15-19(22)18(16)10-7-4-5-8-11-20(23)24/h12-18,21H,2-11H2,1H3,(H,23,24)/b14-12+/t16-,17-,18?/m0/s1
InChI key
BGKHCLZFGPIKKU-BGEXMHJYSA-N
General description
Application
Biochem/physiol Actions
Packaging
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Repr. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service