Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

890701C

Avanti

14:0 EPC (Cl Salt)

Avanti Research - A Croda Brand 890701C

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-ethylphosphocholine (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C38H77NO8PCl
CAS Number:
Molecular Weight:
742.45
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (890701C-10mg)
pkg of 1 × 2.5 mL (890701C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890701C

concentration

10 mg/mL (890701C-10mg)
10 mg/mL (890701C-25mg)

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O)OCC.[Cl-]

Application

14:0 EPC (Cl Salt) is suitable:
  • for combining with a co-lipid, in liposome preparation by sonication[1]
  • for the preparation of cationic lipid mixtures to establish its superior transfection activity[2]
  • for the study of dehydration dynamics[3]

Biochem/physiol Actions

14:0 EPC (Cl Salt) is a cationic lipid. It efficiently mediates gene delivery into the cells.[4] 14:0 EPC (Cl Salt) serves as a non-viral delivery vector.[1]

Packaging

5 mL Clear Glass Sealed Ampule (890701C-10mg)
5 mL Clear Glass Sealed Ampule (890701C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient in vivo delivery of DNA to pulmonary cells using the novel lipid EDMPC
Gorman CM
Gene Therapy, 4(9), 983-983 (1997)
The dehydration dynamics of a model cell membrane induced by cholesterol analogue 6-ketocholestanol investigated using sum frequency generation vibrational spectroscopy
Ma S, et al.
Science China: Chemistry, 58(7), 1176-1186 (2015)
Aspects of nonviral gene therapy: correlation of molecular parameters with lipoplex structure and transfection efficacy in pyridinium-based cationic lipids
Parvizi P, et al.
International Journal of Pharmaceutics, 461(1-2), 145-156 (2014)
Analysis of lipoplex structure and lipid phase changes
Liposomes, 399-423 (2010)
Rumiana Koynova et al.
The journal of physical chemistry. B, 111(27), 7786-7795 (2007-06-19)
Some mixtures of two cationic lipids including phospholipid compounds (O-ethylphosphatidylcholines) as well as common, commercially available cationic lipids, such as dimethylammonium bromides and trimethylammonium propanes, deliver therapeutic DNA considerably more efficiently than do the separate molecules. In an effort to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service