Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880680C

Avanti

C16 PEG750 Ceramide

Avanti Research - A Croda Brand 880680C

Synonym(s):

N-palmitoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)750]}

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880680C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880680C

concentration

10 mg/mL (880680C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCC(OC[C@]([H])(NC(CCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC)=O)OOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC.C=C

General description

Ceramide is a sphingolipid metabolite. The ceramides differ in their acyl chain length.[1]

Application

C16 PEG750 Ceramide might be used as a control to test for nonspecific interactions during titrations of DNA-nanotube complexes with polyethylene glycol (PEG)-conjugated lipids.[2] It is also suitable for use in nanoliposomal formulation.[3][4]

Biochem/physiol Actions

Therapeutic drugs enclosed in poly-ethylene glycol (PEG) conjugated ceramide serves as liposomal carriers, ensuring a longer circulation period. It exhibits a neutral surface electrostatic charge which prevents the leakage of positively charged encapsulated drug molecule.[5]

Packaging

5 mL Clear Glass Sealed Ampule (880680C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Liu et al.
Blood, 116(20), 4192-4201 (2010-07-31)
The natural killer (NK) type of aggressive large granular lymphocytic (LGL) leukemia is a fatal illness that pursues a rapid clinical course. There are no effective therapies for this illness, and pathogenetic mechanisms remain undefined. Here we report that the
Pavan P Adiseshaiah et al.
Cancer letters, 337(2), 254-265 (2013-05-15)
Autophagy, a catabolic survival pathway, is gaining attention as a potential target in cancer. In human liver and colon cancer cells, treatment with an autophagy inducer, nanoliposomal C6-ceramide, in combination with the autophagy maturation inhibitor, vinblastine, synergistically enhanced apoptotic cell
Prakrit V Jena et al.
ACS nano, 11(11), 10689-10703 (2017-09-13)
Lipid accumulation within the lumen of endolysosomal vesicles is observed in various pathologies including atherosclerosis, liver disease, neurological disorders, lysosomal storage disorders, and cancer. Current methods cannot measure lipid flux specifically within the lysosomal lumen of live cells. We developed
M S Webb et al.
Biochimica et biophysica acta, 1372(2), 272-282 (1998-07-24)
Poly(ethylene glycol) (PEG) conjugated lipids have been used to increase the circulation longevity of liposomal carriers encapsulating therapeutic compounds. PEG is typically conjugated to distearoylphosphatidylethanolamine (DSPE) via a carbamate linkage that results in a net negative charge on the phosphate
Michal Levy et al.
IUBMB life, 62(5), 347-356 (2010-03-12)
In mammals, ceramide, a key intermediate in sphingolipid metabolism and an important signaling molecule, is synthesized by a family of six ceramide synthases (CerS), each of which synthesizes ceramides with distinct acyl chain lengths. There are a number of common

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service