Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880610C

Avanti

14:0 PEG750 PE

Avanti Research - A Croda Brand 880610C

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-750] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880610C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880610C

concentration

10 mg/mL (880610C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

General description

16:0 PEG750 PE is a polyethylene glycol (PEG) conjugated phospholipid. The molecular weight and exact mass are averages based on the polydispersity of PEG.

Application

14:0 PEG750 PE is suitable for use as a stabilizing lipid in the preparation of folate receptor-targeting magneto liposomes.[1] It might also be used for graphene surface modification, to study protein translocation.[2]

Biochem/physiol Actions

14:0 PEG750 PE contains a neutral hydrophilic polyethylene glycol (PEG) chain. Thus, it efficiently inhibits protein adsorption during graphene nanopores modification.[2]

Packaging

5 mL Clear Glass Sealed Ampule (880610C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geoffrey D Bothun et al.
Nanomedicine : nanotechnology, biology, and medicine, 7(6), 797-805 (2011-03-23)
Folate-targeted cationic magnetoliposomes (FTMLs) have been prepared with coencapsulated doxorubicin (DOX) and anionic superparamagnetic iron oxide (SPIO) nanoparticles (NPs) with 5 nm γ-Fe(2)O(3) cores and 16 nm hydrodynamic diameters. NP encapsulation (89%) was confirmed by cryogenic transmission electron microscopy (TEM)
Y P Shan et al.
Nanotechnology, 24(49), 495102-495102 (2013-11-16)
Studies of DNA translocation through graphene nanopores have revealed their potential for DNA sequencing. Here we report a study of protein translocation through chemically modified graphene nanopores. A transmission electron microscope (TEM) was used to cut nanopores with diameters between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service