Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880430C

Avanti

18:1 PEG350 PE

Avanti Research - A Croda Brand 880430C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-350] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
51171641
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880430C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880430C

concentration

10 mg/mL (880430C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

Application

18:1 PEG350 PE is suitable for liposome preparation.[1]

Biochem/physiol Actions

Polyethylene glycol (PEG)-lipid conjugates are used extensively to improve circulation times for liposome encapsulated drugs.[2] It promotes sustained injectable delivery system for insulin[3] and superoxide dismutase.[4] Cationic liposomes incorporating PEG-lipid is useful as carriers for oligonucleotides resulting in enhanced uptake by cells.[5]

Packaging

5 mL Clear Glass Sealed Ampule (880430C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cationic liposomes coated with polyethylene glycol as carriers for oligonucleotides
Meyer O, et al.
The Journal of biological chemistry, 273(25), 15621-15627 (1998)
A protocol for the systematic and quantitative measurement of protein?lipid interactions using the liposome-microarray-based assay.
Saliba AE, et al.
Nature Protocols, 11(6), 15621-15627 (2016)
Intravenous administration of superoxide dismutase entrapped in long circulating liposomes: II. In vivo fate in a rat model of adjuvant arthritis
Corvo M, et al.
Biochimica et Biophysica Acta - Biomembranes, 1419(2), 325-334 (1999)
Pharmacodynamics of insulin in polyethylene glycol-coated liposomes
Kim A, et al.
International Journal of Pharmaceutics, 180(1), 75-81 (1999)
Selective transfer of a lipophilic prodrug of 5-fluorodeoxyuridine from immunoliposomes to colon cancer cells
Koning GA, et al.
Biochimica et Biophysica Acta - Biomembranes, 1420(1-2), 153-167 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service