Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880300C

Avanti

16:0 PEG3000 PE

Avanti Research - A Croda Brand 880300C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-3000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880300C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880300C

concentration

10 mg/mL (880300C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCC)

Application

16:0 PEG3000 PE is suitable for the preparation of liposome[1] and advanced drug delivery nano liposomal system.[2]

Biochem/physiol Actions

Phosphoethanolamine serves as a substrate for several phospholipids of the cell membrane. It is associated with the phospholipid turnover. Phosphoethanolamine also prevents tumor progression.[3]

Packaging

5 mL Clear Glass Sealed Ampule (880300C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic phosphoethanolamine induces cell cycle arrest and apoptosis in human breast cancer MCF-7 cells through the mitochondrial pathway
Ferreira AK, et al.
Biomedicine and Pharmacotherapy, 67(6), 481-487 (2013)
Fractal analysis as a complementary approach to predict the stability of drug delivery nano systems in aqueous and biological media: A regulatory proposal or a dream?
Demetzos C and Pippa N
International Journal of Pharmaceutics, 473(1-2), 213-218 (2014)
Comparison of intravenous lipid emulsion, bicarbonate, and tailored liposomes in rabbit clomipramine toxicity
Cave G, et al.
Academic Emergency Medicine : Official Journal of the Society for Academic Emergency Medicine, 20(10), 1076-1079 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service