Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880280C

Avanti

C16 PEG5000 Ceramide

Avanti Research - A Croda Brand 880280C

Synonym(s):

N-palmitoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)5000]}

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211

form

liquid

packaging

pkg of 1 × 2.5 mL (880280C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880280C

concentration

10 mg/mL (880280C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)COC(CCC([(OCH2CH2)113OCH3])=O)=O

General description

C16 PEG5000 Ceramide is categorized as polymers & polymerizable lipids. Ceramides are prominent lipid molecules of stratum corneum. They belong to the group of sphingolipids.[1]

Application

C16 PEG5000 Ceramide might be used to stabilize nanosized bilayer disks[2] and in the preparation of lipid aggregates.[3]

Biochem/physiol Actions

Topical applications containing ceramides is known to repair and replenish the lipid levels in stratum corneum.[1]

Packaging

5 mL Clear Glass Sealed Ampule (880280C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The design and development of anti-aging formulations
Skin Aging Handbook, 291-325 (2009)
Nanosized bilayer disks: attractive model membranes for drug partition studies
Johansson E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1768(6), 1518-1525 (2007)
Melittin-Lipid interaction: A comparative study using liposomes, micelles and bilayerdisks
Lundquist A, et al.
Biochimica et Biophysica Acta - Biomembranes, 1778(10), 2210-2216 (2008)
Hudson P Pace et al.
Analytical chemistry, 90(21), 13065-13072 (2018-10-24)
Over the last two decades, supported lipid bilayers (SLBs) have been extensively used as model systems to study cell membrane structure and function. While SLBs have been traditionally produced from simple lipid mixtures, there has been a recent surge in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service