Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880170C

Avanti

C8 PEG2000 Ceramide

Avanti Research - A Croda Brand 880170C

Synonym(s):

N-octanoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)2000]}

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (880170C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880170C

concentration

10 mg/mL (880170C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCC)=O)COC(CCC(OOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O.C=C

General description

C8 PEG2000 Ceramide, also called as N-octanoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)2000]}, is a polyethylene glycol (PEG)-ylated ceramide. Ceramide-PEGylated lipoplexes are more effective than 1,2-distearoyl-sn-glycero-3-phosphoethanolamine (DSPE)-PEGylated lipoplexes in delivering siRNA to targeted sites.[1]

Application

C8 PEG2000 Ceramide, or N-octanoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)2000]} may be used in preparation of PEGylated lipoplexes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (880170C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Lechanteur et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 93, 493-503 (2016-09-07)
The delivery of small interfering RNA (siRNA) is an attractive therapeutic approach to treat several pathologies, such as viral infections or cancers. However, the stability and the efficacy of these biotherapies are still a major obstacle to their use. Cationic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service