Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

880160C

Avanti

16:0 PEG2000 PE

Avanti Research - A Croda Brand 880160C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211

form

liquid

packaging

pkg of 1 × 2.5 mL (880160C-25mg)
pkg of 2 × 4 mL (880160C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880160C

concentration

10 mg/mL (880160C-25mg)
25 mg/mL (880160C-200mg)

shipped in

dry ice

Storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

General description

16:0 PEG2000 PE, also called as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (DPPEmPEG2000), is a polyethylene glycol (PEG)-modified lipid. Insertion of polyethylene glycol (PEG)-modified lipid to lipid- bilayer helps in formation of sterically stabilized liposome (SSL), which have an ability to spontaneously deliver drug to targeted cells.[1]

Application

16:0 PEG2000 PE, or 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] has been to reduce the adhesion strength between the membrane and the glass during adhesion of cytoskeletal vesicles under hypertonic osmotic stress.[2]

Packaging

5 mL Clear Glass Sealed Ampule (880160C-200mg)
5 mL Clear Glass Sealed Ampule (880160C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renu Maan et al.
Biophysical journal, 115(12), 2395-2402 (2018-11-21)
Regulation of adhesion is a ubiquitous feature of living cells, observed during processes such as motility, antigen recognition, or rigidity sensing. At the molecular scale, a myriad of mechanisms are necessary to recruit and activate the essential proteins, whereas at
Joy Wolfram et al.
Colloids and surfaces. B, Biointerfaces, 114, 294-300 (2013-11-13)
An essential requisite for the design of nanodelivery systems is the ability to characterize the size, homogeneity and zeta potential of nanoparticles. Such properties can be tailored in order to create the most efficient drug delivery platforms. An important question
Kihyeun Kim et al.
Biosensors & bioelectronics, 150, 111885-111885 (2019-11-25)
Solid-phase, single-step biosensors are crucial for the development of portable, reusable, and convenient biosensors, otherwise known as point-of-care (POC) testing. Although high-performance single-step biosensors based on the principle of Förster resonance energy transfer (FRET) and using upconversion nanoparticles (UCNPs) functionalized
D Kirpotin et al.
FEBS letters, 388(2-3), 115-118 (1996-06-17)
Plasma-stable liposomes (100 nm) were prepared from dioleoylphosphatidylethanolamine (DOPE) and 3-6 mol% of a new disulfide-linked poly(ethylene glycol)-phospholipid conjugate (mPEG-DTP-DSPE). In contrast to similar preparations containing non-cleavable PEG-phospholipid conjugate, thiolytic cleavage of the grafted polymer chains facilitated rapid and complete
Ilaria Arduino et al.
International journal of pharmaceutics, 583, 119351-119351 (2020-04-28)
Here, polyethylene glycol (PEG)-stabilized solid lipid nanoparticles (SLNs) containing Pt(IV) prodrugs derived from kiteplatin were designed and proposed as novel nanoformulations potentially useful for the treatment of glioblastoma multiforme. Four different Pt(IV) prodrugs were synthesized, starting from kiteplatin by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service