Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

878120C

Avanti

C18 Lyso PAF

Avanti Research - A Croda Brand 878120C

Synonym(s):

1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H56NO6P
CAS Number:
Molecular Weight:
509.70
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (878120C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878120C

concentration

5 mg/mL (878120C-5mg)

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

Storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCCCCCCCCCCCC

InChI

1S/C26H56NO6P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22-31-24-26(28)25-33-34(29,30)32-23-21-27(2,3)4/h26,28H,5-25H2,1-4H3/t26-/m1/s1

Inchi Key

XKBJVQHMEXMFDZ-AREMUKBSSA-N

General description

C18-lysoPAF or 1-O-octadecyl-2-hydroxy-sn-glycero-3-phosphocholine, is a precursor and metabolite of 1-O-octadecyl-2-acetyl-sn-glycero-3-phosphocholine (C18:0 PAF). Lyso-platelet activating factor (PAF) is synthesized from PAF acetylhydrolases by the degradation of PAF. It is also synthesized from 1-O-alkyl-2-arachidonoyl-sn-glycero-3-phosphocholine (1-alkyl-phosphatidylcholine) by the enzyme phospholipase A2 via the remodeling pathway.[1]

Packaging

5 mL Clear Glass Sealed Ampule (878120C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideo Shindou et al.
The Journal of biological chemistry, 282(9), 6532-6539 (2006-12-22)
Platelet-activating factor (PAF) is a potent proinflammatory lipid mediator eliciting a variety of cellular functions. Lipid mediators, including PAF are produced from membrane phospholipids by enzymatic cascades. Although a G protein-coupled PAF receptor and degradation enzymes have been cloned and
Kimberly D Dyer et al.
Journal of immunology (Baltimore, Md. : 1950), 184(11), 6327-6334 (2010-04-28)
Platelet-activating factor (PAF [1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine]) is a phospholipid mediator released from activated macrophages, mast cells, and basophils that promotes pathophysiologic inflammation. Eosinophil responses to PAF are complex and incompletely elucidated. We show in this article that PAF and its 2-deacetylated metabolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service