Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

872110C

Avanti

18:1 PE-N-18:1

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-oleoyl (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C59H113N2O9P
Molecular Weight:
1025.51
UNSPSC Code:
12352211

assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 10 mg (872110C-10mg)
pkg of 1 × 5 mg (872110C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (872110C-10MG)
5 mg/mL (872110C-5MG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCCNC(CCCCCCC/C=C\CCCCCCCC)=O)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O)=O.[NH4+]

Packaging

5ML Clear Glass Sealed Ampule (872110C-10MG)
5ML Clear Glass Sealed Ampule (872110C-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

target_organs

Liver,Kidney, Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service