Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870738P

Avanti

19:0 Coenzyme A

Avanti Research - A Croda Brand 870738P, powder

Synonym(s):

nonadecanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H81N10O17P3S
CAS Number:
Molecular Weight:
1099.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870738P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870738P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C40H72N7O17P3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-31(49)68-24-23-42-30(48)21-22-43-38(52)35(51)40(2,3)26-61-67(58,59)64-66(56,57)60-25-29-34(63-65(53,54)55)33(50)39(62-29)47-28-46-32-36(41)44-27-45-37(32)47/h27-29,33-35,39,50-51H,4-26H2,1-3H3,(H,42,48)(H,43,52)(H,56,57)(H,58,59)(H2,41,44,45)(H2,53,54,55)

InChI key

YUJZGRUJMWKAJF-UHFFFAOYSA-N

General description

19:0 Coenzyme A, also known as nonadecanoyl coenzyme A, is a coenzyme A derivative of nonadecanoic acid.

Packaging

5 mL Amber Glass Screw Cap Vial (870738P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena Topolska et al.
Biomolecules, 10(1) (2020-01-16)
De novo fatty acid synthesis is a pivotal enzymatic process in all eukaryotic organisms. It is involved in the conversion of glucose and other nutrients to fatty acyl (FA) chains, that cells use as building blocks for membranes, energy storage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service