Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870601P

Avanti

Azelaoyl-PAF

Avanti Research - A Croda Brand 870601P, powder

Synonym(s):

1-hexadecyl-2-azelaoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H66NO9P
CAS Number:
Molecular Weight:
651.85
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (870601P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870601P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCC(O)=O)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C33H66NO9P/c1-5-6-7-8-9-10-11-12-13-14-15-16-20-23-27-40-29-31(30-42-44(38,39)41-28-26-34(2,3)4)43-33(37)25-22-19-17-18-21-24-32(35)36/h31H,5-30H2,1-4H3,(H-,35,36,38,39)/t31-/m1/s1

InChI key

ZDFOCDTXDPKJKA-WJOKGBTCSA-N

Application

Azelaoyl-PAF may be used to study its binding affinity with 4F or 2F anti-inflammatory peptides.[1]

Biochem/physiol Actions

Azelaoyl-PAF (1-hexadecyl-2-azelaoyl-sn-glycero-3-phosphocholine) is a low molecular weight alkyl phosphatidylcholine. It is a constituent of the lipid pool within oxidized low-density lipoprotein (oxLDL) particles. Azelaoyl-PAF serves as a high affinity ligand and agonist for peroxisome proliferator-activated receptor-γ (PPARγ).

Packaging

5 mL Clear Glass Sealed Ampule (870601P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel F Epand et al.
Biochimica et biophysica acta, 1788(9), 1967-1975 (2009-06-30)
The peptide 4F is known to have potent anti-atherogenic activity. 4F is an 18 residue peptide that has a sequence capable of forming a class A amphipathic helix. Several other class A amphipathic helical, 18 residue peptides with the same
S S Davies et al.
The Journal of biological chemistry, 276(19), 16015-16023 (2001-03-30)
Synthetic high affinity peroxisome proliferator-activated receptor (PPAR) agonists are known, but biologic ligands are of low affinity. Oxidized low density lipoprotein (oxLDL) is inflammatory and signals through PPARs. We showed, by phospholipase A(1) digestion, that PPARgamma agonists in oxLDL arise
P Tontonoz et al.
Cell, 93(2), 241-252 (1998-05-06)
The formation of foam cells from macrophages in the arterial wall is characterized by dramatic changes in lipid metabolism, including increased expression of scavenger receptors and the uptake of oxidized low-density lipoprotein (oxLDL). We demonstrate here that the nuclear receptor
Douglas S Watson et al.
Vaccine, 27(34), 4672-4683 (2009-06-13)
The membrane proximal region (MPR) of HIV-1 gp41 is a desirable target for development of a vaccine that elicits neutralizing antibodies since the patient-derived monoclonal antibodies, 2F5 and 4E10, bind to the MPR and neutralize primary HIV isolates. The 2F5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service