Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870436P

Avanti

C16:0 anandamide phosphate

Avanti Research - A Croda Brand 870436P, powder

Synonym(s):

palmitoyl ethanolamide phosphate (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37NO5PNa
Molecular Weight:
401.45
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (870436P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870436P

shipped in

dry ice

Storage temp.

−20°C

SMILES string

O=C(CCCCCCCCCCCCCCC)NCCOP(O)([O-])=O.[Na+]

General description

C16:0 anandamide is fatty acid amide, endogenic in nature. It is a member of the N-acylethanolamines family. Palmitoyl ethanolamide is present in egg yolk and peanut.
C16:0 anandamide phosphate is a phosphate ester of C16:0 anandamide. C16:0 anandamide, also known as N-palmitoyl ethanolamide, is a N-acyl ethanolamide derivative of palmitic acid.

Biochem/physiol Actions

Anandamide is a ligand for cannabinoid receptors. Anandamide possesses anti-inflammatory and pain killing action. It is effective in relieving pain associated with nerve compression syndrome and pudendal neuralgia. Anandamide is also useful in treating atopic eczema. Its effect is mediated by cannabinoid CB1 and CB2 receptors.
C16:0 anandamide or palmitoyl ethanolamide exhibits anti-inflammatory and analgesic properties. It is currently used as a veterinary anti-inflammatory agent.

Packaging

5 mL Amber Glass Screw Cap Vial (870436P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palmitoylethanolamide ameliorates carbon tetrachloride-induced liver fibrosis in rats
Ohara M, et al.
Frontiers in Pharmacology, 9, 698-711 (2018)
Ocular hypotensive effect of oral palmitoyl-ethanolamide: a clinical trial
Gagliano C, et al.
Investigative Ophthalmology & Visual Science, 52(9), 6096-6100 (2011)
Protective role of palmitoylethanolamide in contact allergic dermatitis
Petrosino S, et al.
Allergy, 65(6), 698-711 (2010)
Daria Vinokurova et al.
Frontiers in pharmacology, 9, 698-698 (2018-07-19)
Epipial application is one of the approaches for drug delivery into the cortex. However, passive diffusion of epipially applied drugs through the cortical depth may be slow, and different drug concentrations may be achieved at different rates across the cortical
Natsuo Ueda et al.
Biochimica et biophysica acta, 1801(12), 1274-1285 (2010-08-26)
Ethanolamides of different long-chain fatty acids constitute a class of endogenous lipid molecules generally called N-acylethanolamines (NAEs). They contain N-arachidonoylethanolamine (anandamide), N-palmitoylethanolamine, and N-oleoylethanolamine, which receive considerable attention because of their actions as an endogenous cannabinoid receptor ligand (endocannabinoid), an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service