Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870412C

Avanti

16:0 DG Galloyl

Avanti Research - A Croda Brand 870412C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-galloyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H72O9
CAS Number:
Molecular Weight:
721.02
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870412C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870412C

concentration

10 mg/mL (870412C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COC(C1=CC(O)=C(O)C(O)=C1)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C42H72O9/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-39(45)49-33-36(34-50-42(48)35-31-37(43)41(47)38(44)32-35)51-40(46)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h31-32,36,43-44,47H,3-30,33-34H2,1-2H3/t36-/m1/s1

InChI key

VUMJZTCVWBBKRP-PSXMRANNSA-N

Application

16:0 DG Galloyl or 1,2-dipalmitoyl-sn-glycero-3-galloyl (DPGG), is used as a phenolic lipid to study the surface functionalization of metal-organic frameworks (MOF) particles.

Biochem/physiol Actions

16:0 DG Galloyl, also known as 1,2-dipalmitoyl-sn-glycero-3-galloyl (DPGG), is a phenolic-based lipid molecule. It has an ability to form phenolic metal complexes with lead (II) ions.

Packaging

5 mL Clear Glass Sealed Ampule (870412C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile Surface Functionalization of Metal--Organic Frameworks through Direct Metal Coordination with a Phenolic Lipid Enables Diverse Applications
Zhu W, et al.
Advances in Functional Materials, 28(16), 1705274-1705274 (2018)
Janus particles for (bio) sensing
Yanez-Sedeno P, et al.
Applied Materials Today, 9, 276-288 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service