Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870314O

Avanti

18:1 DGS

Avanti Research - A Croda Brand 870314O

Synonym(s):

1,2-dioleoyl-sn-glycero-3-succinate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H76O8
CAS Number:
Molecular Weight:
721.06
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 2 × 100 mg (870314O-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870314O

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C43H76O8/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(46)49-37-39(38-50-42(47)36-35-40(44)45)51-43(48)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,39H,3-16,21-38H2,1-2H3,(H,44,45)/b19-17-,20-18-/t39-/m1/s1

InChI key

VZGIZLLBNVAMQT-NYVOMTAGSA-N

General description

18:1 DGS, also known as 1,2-dioleoyl-sn-glycero-3-succinate (DOSG), is a pH sensitive non-phospholipid.[1] It comprises two oleoyl chains and both the chains are involved in forming stable fluidic bilayers.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870314O-200mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unique supramolecular assembly of a redox protein with nucleic acids onto hybrid bilayer: towards a dynamic DNA chip
Boireau W, et al.
Biosensors And Bioelectronics, 20(8), 1631-1637 (2005)
Lipid-dependent gating of a voltage-gated potassium channel
Zheng H, et al.
Nature Communications, 2, 250-250 (2011)
W Boireau et al.
Biosensors & bioelectronics, 20(8), 1631-1637 (2005-01-01)
Highly controlled supramolecular assemblies combining a genetically engineered redox protein, cytochrome b5, and modified oligonucleotides are presented. Modified b5 and DNA are covalently assembled through a hetero bifunctional cross-linker to give a unique hybrid molecular species. Moreover, the assembly includes
Hui Zheng et al.
Nature communications, 2, 250-250 (2011-03-24)
Recent studies hypothesized that phospholipids stabilize two voltage-sensing arginine residues of certain voltage-gated potassium channels in activated conformations. It remains unclear how lipids directly affect these channels. Here, by examining the conformations of the KvAP in different lipids, we showed
Sungsoo Lee et al.
Journal of visualized experiments : JoVE, (77), e50436-e50436 (2013-07-31)
To study the lipid-protein interaction in a reductionistic fashion, it is necessary to incorporate the membrane proteins into membranes of well-defined lipid composition. We are studying the lipid-dependent gating effects in a prototype voltage-gated potassium (Kv) channel, and have worked

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service