Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870312C

Avanti

16:0 DGS

Avanti Research - A Croda Brand 870312C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-succinate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C39H72O8
CAS Number:
Molecular Weight:
668.98
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870312C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870312C

concentration

10 mg/mL (870312C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COC(CCC(O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H72O8/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-37(42)45-33-35(34-46-38(43)32-31-36(40)41)47-39(44)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35H,3-34H2,1-2H3,(H,40,41)/t35-/m1/s1

InChI key

GNENAKXIVCYCIZ-PGUFJCEWSA-N

Application

16:0 DGS has been used in the preparation of liposomes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (870312C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knut-Egil Løkling et al.
Magnetic resonance in medicine, 51(4), 688-696 (2004-04-06)
The use of paramagnetic pH-sensitive liposomes was recently suggested as a new approach for monitoring pathologic changes in pH by MRI. Such liposomes must be stable in blood and selectively release the encapsulated paramagnetic agent when exposed to lower pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service