870307P
Avanti
16:0 rac PC-d80
powder
Synonym(s):
1,2-dipalmitoyl-rac-glycero-3-phosphocholine (d80)
Select a Size
Select a Size
About This Item
assay
99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (870307P-1MG)
pkg of 1 × 5 mg (870307P-5MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand (870307P)
shipped in
dry ice
Storage temp.
−20°C
SMILES string
[O-]P(OC([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])(OC([2H])([2H])C([2H])(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H]
General description
Packaging
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service