Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870265P

Avanti

16:0 Dodecanoyl PE

Avanti Research - A Croda Brand 870265P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H93NO11PNa
CAS Number:
Molecular Weight:
926.23
UNSPSC Code:
12352211

form

powder

packaging

pkg of 1 × 25 mg (870265P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870265P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCCCCCCCC(O)=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C49H94NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-27-31-35-39-48(54)58-43-45(61-49(55)40-36-32-28-22-20-18-16-14-12-10-8-6-4-2)44-60-62(56,57)59-42-41-50-46(51)37-33-29-25-23-24-26-30-34-38-47(52)53;/h45H,3-44H2,1-2H3,(H,50,51)(H,52,53)(H,56,57);/q;+1/p-1/t45-;/m1./s1

InChI key

CUCCNZVUGZZGNC-NZEIQPSFSA-M

General description

16:0 Dodecanoyl PE, also called as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl), belongs to the group of head group modified functionalized lipids.

Application

16:0 Dodecanoyl PE or 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl) is suitable for charged functionalized liposomes preparation.

Packaging

5 mL Clear Glass Sealed Ampule (870265P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ehsan Suleiman et al.
Pharmaceutics, 11(11) (2019-11-23)
Since the first use of liposomes as carriers for antigens, much work has been done to elucidate the mechanisms involved in the encapsulation of vaccine-relevant biomolecules. However, only a few studies have specifically investigated the encapsulation of hydrophilic, non-conformational peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service