Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870265C

Avanti

16:0 Dodecanoyl PE

Avanti Research - A Croda Brand 870265C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C49H93NO11PNa
CAS Number:
Molecular Weight:
926.23
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870265C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870265C

concentration

10 mg/mL (870265C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCCCCCCCC(O)=O)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C49H94NO11P.Na/c1-3-5-7-9-11-13-15-17-19-21-27-31-35-39-48(54)58-43-45(61-49(55)40-36-32-28-22-20-18-16-14-12-10-8-6-4-2)44-60-62(56,57)59-42-41-50-46(51)37-33-29-25-23-24-26-30-34-38-47(52)53;/h45H,3-44H2,1-2H3,(H,50,51)(H,52,53)(H,56,57);/q;+1/p-1/t45-;/m1./s1

InChI key

CUCCNZVUGZZGNC-NZEIQPSFSA-M

General description

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl) (16:0 Dodecanoyl PE) is a head group modified functionalized lipid.

Application

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanoyl) (16:0 Dodecanoyl PE) may be used in preparation of charged functionalized liposomes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (870265C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ehsan Suleiman et al.
Pharmaceutics, 11(11) (2019-11-23)
Since the first use of liposomes as carriers for antigens, much work has been done to elucidate the mechanisms involved in the encapsulation of vaccine-relevant biomolecules. However, only a few studies have specifically investigated the encapsulation of hydrophilic, non-conformational peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service