Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870205C

Avanti

16:0 PDP PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio)propionate] (sodium salt), chloroform

Synonym(s):

Sodium (2R)-2,3-bis(hexadecanoyloxy)propyl 2-{3-[(pyridin-2-yl)disulfanyl]propanamido}ethyl phosphate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C45H80N2O9PNaS2
CAS Number:
Molecular Weight:
911.22
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870205C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870205C

concentration

10 mg/mL (870205C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCSSC1=CC=CC=N1)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C45H81N2O9PS2.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-32-44(49)53-39-41(56-45(50)33-28-26-24-22-20-18-16-14-12-10-8-6-4-2)40-55-57(51,52)54-37-36-46-42(48)34-38-58-59-43-31-29-30-35-47-43;/h29-31,35,41H,3-28,32-34,36-40H2,1-2H3,(H,46,48)(H,51,52);/q;+1/p-1/t41-;/m1./s1

InChI key

IUSHQKJFHVODPF-QUEDAWNRSA-M

General description

16:0 PDP PE, also known as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio) propionate, is a head group modified functionalized lipid. Functionalized lipids mainly act as reporting molecules.[1]

Application

16:0 PDP PE has been used as an inner lipid to synthesizehigh density lipoprotein-like nanoparticles (HDL-like NPs).[2]
16:0 PDP PE has been used to synthesize high-density lipoprotein (HDL)-like nano particles.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870205C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic high-density lipoprotein-like nanoparticles potently inhibit cell signaling and production of inflammatory mediators induced by lipopolysaccharide binding Toll-like receptor 4
Foit L and Thaxton CS
Biomaterials, 100, 67-75 (2016)
Linda Foit et al.
Biomaterials, 100, 67-75 (2016-06-01)
Toll-like receptor 4 (TLR4) plays a critical role in the innate immune system. Stimulation of TLR4 occurs upon binding lipopolysaccharide (LPS), a component of Gram-negative bacterial cell walls. Due to the potency of the induced inflammatory response, there is a
Synthetic high-density lipoprotein-like nanoparticles potently inhibit cell signaling and production of inflammatory mediators induced by lipopolysaccharide binding Toll-like receptor 4
Foit L and Thaxton CS
Biomaterials, 100, 67-75 (2016)
Functionalized lipids and surfactants for specific applications
Kepczynski M and Rog T
Biochimica et Biophysica Acta - Biomembranes, 1858(10), 2362-2379 (2016)
Nicholas L Angeloni et al.
Scientific reports, 6, 22915-22915 (2016-03-12)
Exosomes are produced by cells to mediate intercellular communication, and have been shown to perpetuate diseases, including cancer. New tools are needed to understand exosome biology, detect exosomes from specific cell types in complex biological media, and to modify exosomes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service