Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870142P

Avanti

18:1 Dodecanylamine PE

Avanti Research - A Croda Brand 870142P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C53H101N2O9P
CAS Number:
Molecular Weight:
941.35
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870142P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870142P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN(C(CCCCCCCCCCC[NH3+])=O)[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C53H101N2O9P/c1-3-5-7-9-11-13-15-17-19-21-23-27-31-35-39-43-52(57)61-48-50(64-53(58)44-40-36-32-28-24-22-20-18-16-14-12-10-8-6-4-2)49-63-65(59,60)62-47-46-55-51(56)42-38-34-30-26-25-29-33-37-41-45-54/h17-20,50H,3-16,21-49,54H2,1-2H3,(H,55,56)(H,59,60)/b19-17-,20-18-/t50-/m1/s1

InChI key

HOOLPMPNZPBWNI-WSPRLCGESA-N

General description

18:1 Dodecanylamine PE, also called as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio)propionate], belongs to the class of head group modified functionalized lipids. Functionalized lipids generally perform the role of reporter molecules.[1]

Packaging

5 mL Clear Glass Sealed Ampule (870142P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalized lipids and surfactants for specific applications
Kepczynski M and Rog T
Biochimica et Biophysica Acta - Biomembranes, 1858(10), 2362-2379 (2016)
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service