Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870140C

Avanti

16:0 Dodecanylamine PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine), chloroform

Synonym(s):

2-(12-azaniumyldodecanoylamino)ethyl[(2R)-2,3 di(hexadecanoyloxy)propyl] phosphate

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C49H97N2O9P
CAS Number:
Molecular Weight:
889.28
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870140C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870140C

concentration

10 mg/mL (870140C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN([H])C(CCCCCCCCCCC[NH3+])=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C49H97N2O9P/c1-3-5-7-9-11-13-15-17-19-23-27-31-35-39-48(53)57-44-46(60-49(54)40-36-32-28-24-20-18-16-14-12-10-8-6-4-2)45-59-61(55,56)58-43-42-51-47(52)38-34-30-26-22-21-25-29-33-37-41-50/h46H,3-45,50H2,1-2H3,(H,51,52)(H,55,56)/t46-/m1/s1

InChI key

MFAZBSZBKVFZAN-YACUFSJGSA-N

General description

16:0 Dodecanylamine PE, also referred as 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(dodecanylamine), is a head group modified functionalized lipid. Functionalized lipids functions as reporter molecules.[1]

Application

16:0 Dodecanylamine PE has been used as a co-immobilized lipid to stabilize the haem group of Cyt c.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870140C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functionalized lipids and surfactants for specific applications
Kepczynski M and Rog T
Biochimica et Biophysica Acta - Biomembranes, 1858(10), 2362-2379 (2016)
Md Aminur Rahman et al.
Analytical chemistry, 84(15), 6654-6660 (2012-07-19)
Enhanced stability and sensitivity of a superoxide anion radical (O(2)(•-)) microbiosensor were achieved through the sequential immobilization of lipid and cytochrome c (Cyt c) covalently bonded onto a conducting polymer layer that showed a clear quasi-reversible direct electron transfer (DET)
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service