Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

870122C

Avanti

18:1 Caproylamine PE

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(hexanoylamine)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C47H89N2O9P
CAS Number:
Molecular Weight:
857.19
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 2.5 mL (870122C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (870122C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCN(C(CCCCC[NH3+])=O)[H])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C47H89N2O9P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-33-37-46(51)55-42-44(43-57-59(53,54)56-41-40-49-45(50)36-32-31-35-39-48)58-47(52)38-34-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,44H,3-16,21-43,48H2,1-2H3,(H,49,50)(H,53,54)/b19-17-,20-18-/t44-/m1/s1

InChI key

OVCGTEBXOCFVEO-OHRCZFALSA-N

General description

18:1 Caproylamine PE is a head group modified functionalized lipid. Functionalized lipids mainly act as reporting molecules.[1]

Application

18:1 Caproylamine PE may be used as a positive control in competitive enzyme-linked immunosorbent assay (ELISA) for measuring affinities between glycolipid analogs and human or mouse CD1d molecules.[2] It has also been used as a lipid antigen to generate CD1d-Ig–based artificial antigen-presenting cells (aAPC).[3]

Packaging

5 mL Clear Glass Sealed Ampule (870122C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tonya J Webb et al.
Cancer research, 72(15), 3744-3752 (2012-06-01)
Tumors often display mechanisms to avoid or suppress immune recognition. One such mechanism is the shedding of gangliosides into the local tumor microenvironment, and a high concentration of circulating gangliosides is associated with poor prognosis. In this study, we identify
Xiangming Li et al.
Journal of immunology (Baltimore, Md. : 1950), 183(7), 4415-4421 (2009-09-08)
C-glycoside analogues of alpha-galactosylceramide were shown to activate both human and mouse invariant NKT (iNKT) cells. Among these analogues, GCK152, which has an aromatic ring in the acyl chain, exhibited a stronger stimulatory activity against human iNKT cells and a
Mariusz Kepczynski et al.
Biochimica et biophysica acta, 1858(10), 2362-2379 (2016-03-08)
Synthetic lipids and surfactants that do not exist in biological systems have been used for the last few decades in both basic and applied science. The most notable applications for synthetic lipids and surfactants are drug delivery, gene transfection, as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service