Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860730P

Avanti

C16 Galactosyl (a) Dihydroceramide (d18:0/16:0)

Avanti Research - A Croda Brand, powder

Synonym(s):

D-galactosyl-a-1,1′ N-palmitoyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C40H79NO8
CAS Number:
Molecular Weight:
702.06
UNSPSC Code:
12352211
NACRES:
NA.25

assay

99% (TLC)

form

powder

packaging

1 ea of 5 mg (860730P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@]([C@@]([H])(CCCCCCCCCCCCCCC)O)(NC(CCCCCCCCCCCCCCC)=O)CO[C@@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI key

BLGKYYVFGMKTEZ-SGUSGNBCSA-N

Packaging

5 mL Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service