860647P
Avanti
C18:1 Ceramide (d17:1/18:1(9Z))
Avanti Research™ - A Croda Brand 860647P, powder
Synonym(s):
N-oleoyl-D-erythro-sphingosine (C17 base)
Select a Size
Select a Size
About This Item
form
powder
packaging
pkg of 1 × 10 mg (860647P-10mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 860647P
shipped in
dry ice
storage temp.
−20°C
SMILES string
OC[C@]([H])(NC(CCCCCCC/C=C\CCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCC
General description
Application
Biochem/physiol Actions
Packaging
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
No data available
flash_point_c
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service