Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860644P

Avanti

C2 Ceramide (d17:1/2:0)

Avanti Research - A Croda Brand 860644P, powder

Synonym(s):

N-acetoyl-D-erythro-sphingosine (C17 base)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H37NO3
CAS Number:
Molecular Weight:
327.50
UNSPSC Code:
12352100

form

powder

packaging

pkg of 1 × 10 mg (860644P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860644P

lipid type

sphingolipids
deuterated lipids

shipped in

dry ice

Storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(C)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCC

General description

C2 Ceramide (N-acetoyl-D-erythro-sphingosine) is a synthetic ceramide analog.[1] It is cell-permeable in nature.[1][2] C2 Ceramide belongs to the class of short chain ceramides, also known as the soluble swelling amphiphiles.[3] It is widely present in foods ranging from eggs to soybeans.[4]

Application

C2 Ceramide (d17:1/2:0) has been used as an internal standard in tandem mass spectrometry for sphingolipid analysis.[5]

Biochem/physiol Actions

C2 Ceramide (C2-Cer) has an ability to mediate internucleosomal DNA fragmentation.[1] It is implicated in steroidogenic pathway.[2] C2 Ceramide has a potential to transform edible oil in to self-standing organogels without any saturated or trans fats supplements.[4] Short chain ceramides will remain intact with liquid-crystalline phospholipids and they facilitate the formation of positive curvature in lipid membranes. These ceramides are usually used as a substitutes for long-chain Cer in various biological experimentation with living cells.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860644P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raft domain reorganization driven by short-and long-chain ceramide: a combined AFM and FCS study
Chiantia S, et al.
Langmuir, 23(14), 7659-7665 (2007)
A potential bioactive hard-stock fat replacer comprised of a molecular gel
Rogers MA, et al.
Food Science & Nutrition, 5(3), 579-587 (2017)
Programmed cell death induced by ceramide
Obeid LM, et al.
Science (New York, N.Y.), 259(5102), 1769-1771 (1993)
The interplay between bioactive sphingolipids and steroid hormones
Lucki NC and Sewer MB
Steroids, 75(6), 390-399 (2010)
Sexual dimorphism in activation of placental autophagy in obese women with evidence for fetal programming from a placenta-specific mouse model
Muralimanoharan S, et al.
Autophagy, 12(5), 752-769 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service