Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860643P

Avanti

Trimethyl Sphingosine (d17:1)

Avanti Research - A Croda Brand, powder

Synonym(s):

N,N,N-trimethyl-D-erythro-sphingosine (C17 base) (methyl sulfate salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H45NO6S
CAS Number:
Molecular Weight:
439.65
UNSPSC Code:
12352211

assay

99% (TLC)

form

powder

packaging

1 ea of 5 mg (860643P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@]([N+](C)(C)C)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCC.O=S([O-])(OC)=O

General description

Trimethyl Sphingosine is a derivative of sphingosine.[1]

Packaging

5 mL Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential signaling of sphingosine derivatives in U937 human monocytes depends on the degree of N-methylation
Kim H L, et al.
Prostaglandins & other lipid mediators, 68-72 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service