Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860621P

Avanti

Dimethyl Sphinganine (d18:0)

Avanti Research - A Croda Brand, powder

Synonym(s):

N,N-dimethyl-D-erythro-dihydrosphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H43NO2
Molecular Weight:
329.56
UNSPSC Code:
12352211
NACRES:
NA.25

assay

99% (TLC)

form

powder

packaging

1 ea of 5 mg (860621P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N(C)C)([H])[C@]([H])(O)CCCCCCCCCCCCCCC

Packaging

5 mL Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service