Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860581C

Avanti

18:1-12:0 Biotin PG

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl-2-[12-biotinyl(aminododecanoyl)]-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H83 N3NaO12PS
Molecular Weight:
956.19
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (860581C-10mg)
pkg of 1 × 1 mL (860581C-1mg)
pkg of 1 × 1 mL (860581C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (860581C-1mg)
10 mg/mL (860581C-10mg)
5 mg/mL (860581C-5mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

Application

18:1-12:0 Biotin PG or 1-oleoyl-2-[12-biotinyl(aminododecanoyl)]-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) has been used in lipid blot analysis to study Niemann-Pick C2 (NPC2)-lipid interactions.

Packaging

5 mL Clear Glass Sealed Ampule (860581C-10mg)
5 mL Clear Glass Sealed Ampule (860581C-1mg)
5 mL Clear Glass Sealed Ampule (860581C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leslie A McCauliff et al.
eLife, 8 (2019-10-04)
Unesterified cholesterol accumulation in the late endosomal/lysosomal (LE/LY) compartment is the cellular hallmark of Niemann-Pick C (NPC) disease, caused by defects in the genes encoding NPC1 or NPC2. We previously reported the dramatic stimulation of NPC2 cholesterol transport rates to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service