Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860561P

Avanti

18:1-12:0 Biotin PA

Avanti Research - A Croda Brand 860561P, powder

Synonym(s):

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H81N4O10PS
CAS Number:
Molecular Weight:
877.16
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860561P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860561P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C43H78N3O10PS.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-23-30-40(48)54-33-36(34-55-57(51,52)53)56-41(49)31-24-21-18-15-13-16-19-22-27-32-44-39(47)29-26-25-28-38-42-37(35-58-38)45-43(50)46-42;/h9-10,36-38,42H,2-8,11-35H2,1H3,(H,44,47)(H2,45,46,50)(H2,51,

InChI key

LAYLPTJRIODVKN-MMGYSICESA-N

General description

18:1-12:0 Biotin PA is a biotinylated form of phosphatidic acid, which can be immobilized on a solid support via streptavidin tether.

Application

18:1-12:0 Biotin PA is suitable for use in lipid blot analysis to study Niemann-Pick C2 (NPC2)-lipid interactions.[1]

Packaging

5 mL Clear Glass Sealed Ampule (860561P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

11 - Combustible Solids

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intracellular cholesterol trafficking is dependent upon NPC2 interaction with lysobisphosphatidic acid
McCauliff LA, et al.
eLife, 8 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service