Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860496C

Avanti

Dimethyl Sphingosine (d18:1)

Avanti Research - A Croda Brand 860496C

Synonym(s):

N,N-dimethyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H41NO2
CAS Number:
Molecular Weight:
327.55
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (860496C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860496C

concentration

5 mg/mL (860496C-5mg)

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N(C)C)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C20H41NO2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)19(18-22)21(2)3/h16-17,19-20,22-23H,4-15,18H2,1-3H3/b17-16+/t19-,20+/m0/s1

InChI key

YRXOQXUDKDCXME-YIVRLKKSSA-N

Packaging

5 mL Clear Glass Sealed Ampule (860496C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyo-Lim Kim et al.
Prostaglandins & other lipid mediators, 86(1-4), 18-25 (2008-03-05)
In the present study, we studied N,N-dimethyl-D-erythro-sphingosine (DMS)-induced cell death and its signaling mechanism in U937 human monocytes. We found that DMS induced cell death in a concentration-dependent manner, while sphingosine 1-phosphate did not. DMS also induced DNA fragmentation, nuclear
Juliana Fraga Vasconcelos et al.
Scientific reports, 7(1), 6171-6171 (2017-07-25)
Chagas disease cardiomyopathy is a parasite-driven inflammatory disease to which there are no effective treatments. Here we evaluated the therapeutic potential of N,N-dimethylsphingosine(DMS), which blocks the production of sphingosine-1-phosphate(S1P), a mediator of cellular events during inflammatory responses, in a model
Hyo-Lim Kim et al.
Prostaglandins & other lipid mediators, 86(1-4), 68-72 (2008-05-10)
Previously, we studied N,N-dimethyl-D-erythro-sphingosine (DMS)-induced cell death and signaling in U937 human monocytes; we found that DMS-induced sphingosine kinase- and PKC-independent apoptosis. In the present study, we studied apoptotic responses by three N-methyl derivatives of sphingosine: N-monomethyl-D-erythro-sphingosine (MMS), N,N,N-trimethyl-D-erythro-sphingosine (TMS)
Ji-Yeong Jo et al.
Journal of pharmacological sciences, 107(3), 303-307 (2008-07-19)
Calcium is a ubiquitous second messenger that controls a broad range of cellular functions, and store-operated calcium entry (SOCE) is the primary mechanism of regulated Ca(2+) entry in non-excitable immunocytes. In this study, we found that N,N-dimethyl-D-erythro-sphingosine (DMS) inhibited SOCE.
Yup-Kyung Lee et al.
Molecules and cells, 23(1), 11-16 (2007-04-28)
N,N-dimethyl-D-erythro-sphingosine (DMS) is an N-methyl derivative of sphingosine and an inhibitor of protein kinase C (PKC) and sphingosine kinase (SK). In the present study, we examined the effects of DMS on intracellular Ca2+ concentration, pH, and glutamate uptake in human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service