Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860494C

Avanti

Sphingosine-1-Phosphate (DMA Adduct)

Avanti Research - A Croda Brand 860494C

Synonym(s):

D-erythro-sphingosine-1-phosphate (DMA Adduct)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H45N2O5P
CAS Number:
Molecular Weight:
424.56
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (860494C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860494C

concentration

1 mg/mL (860494C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@]([NH3+])([H])COP([O-])(O)=O.CNC

InChI

1S/C18H38NO5P.C2H7N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(20)17(19)16-24-25(21,22)23;1-3-2/h14-15,17-18,20H,2-13,16,19H2,1H3,(H2,21,22,23);3H,1-2H3/b15-14+;/t17-,18+;/m0./s1

InChI key

WAZPRRNLZCUNSS-ZNWYJMOFSA-N

General description

Sphingosine-1-phosphate (S1P) is a sphingolipid metabolite present in yeast, plants and mammals.[1] It is secreted by endothelial cells.[2]

Biochem/physiol Actions

Sphingosine 1-phosphate (S1P) is a signaling lipid,[1] which is involved in various physiological and pathophysiological processes.[3] It functions as a high-affinity ligand for five sphingosine-1-phosphate receptors (S1P1-S1P5).[3][1] S1P modulates the functions of skin dendritic cells and keratinocytes.[2] It is involved in the pathogenesis of diabetes osteoporosis, atherosclerosis and cancer.[4] S1P plays an important role in maintaining the circulation of T lymphocytes between tissue, plasma and lymph.[2]

Packaging

5 mL Clear Glass Sealed Ampule (860494C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiang Chen et al.
Current cancer drug targets, 19(5), 417-427 (2018-05-02)
Elevated production of the pro-inflammatory cytokine interleukin-6 (IL-6) and dysfunction of IL-6 signaling promotes tumorigenesis and are associated with poor survival outcomes in multiple cancer types. Recent studies showed that the IL-6/GP130/STAT3 signaling pathway plays a pivotal role in pancreatic
Sarah Spiegel et al.
Nature reviews. Molecular cell biology, 4(5), 397-407 (2003-05-03)
The evolutionarily conserved actions of the sphingolipid metabolite, sphingosine-1-phosphate (S1P), in yeast, plants and mammals have shown that it has important functions. In higher eukaryotes, S1P is the ligand for a family of five G-protein-coupled receptors. These S1P receptors are
William J Valentine et al.
Methods in molecular biology (Clifton, N.J.), 874, 77-87 (2012-04-25)
Intracellular Ca(2+) mobilization is a useful readout to screen for agonists or antagonists of G-protein -coupled receptors (GPCRs). Here, we describe methods to conduct high-throughput screening of stably or transiently transfected HTC4 cells expressing the individual S1P1-5 receptor subtypes. The
Michael Maceyka et al.
Trends in cell biology, 22(1), 50-60 (2011-10-18)
The bioactive sphingolipid metabolite sphingosine-1-phosphate (S1P) is now recognized as a critical regulator of many physiological and pathophysiological processes, including cancer, atherosclerosis, diabetes and osteoporosis. S1P is produced in cells by two sphingosine kinase isoenzymes, SphK1 and SphK2. Many cells
C Terranova et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 52, 125-131 (2013-11-13)
In this study we investigate in in vitro myometrial tissue samples of pregnant women: (a) the effects of proton pomp inhibitors (PPIs) (omeprazole, esomeprazole, pantoprazole, lansoprazole and rabeprazole) on spontaneous contractions; (b) the muscle-relaxant efficacy of the most active PPI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service