Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860368C

Avanti

18:0 PC-d83

Avanti Research - A Croda Brand 860368C

Synonym(s):

1,2-distearoyl-d70-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C44H5NO8PD83
CAS Number:
Molecular Weight:
873.66
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860368C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860368C

concentration

10 mg/mL (860368C-10mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H]

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1/i1D3,2D3,3D3,4D3,5D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2,32D2,33D2,34D2,35D2,36D2,37D2,38D2,39D2

InChI key

NRJAVPSFFCBXDT-OKUWEUPRSA-N

General description

18:0 PC-d83 is a deuterated phosphocholine (PC) in which 83 protons of distearoyl are replaced by deuterium.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

18:0 PC-d83 may be used in the preparation of lipid aggregates.[1]

Packaging

5 mL Clear Glass Sealed Ampule (860368C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melittin-Lipid interaction: A comparative study using liposomes, micelles and bilayerdisks
Lundquist A, et al.
Biochimica et Biophysica Acta - Biomembranes, 1778(10), 2210-2216 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service