Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860365C

Avanti

18:0 PC-d70

Avanti Research - A Croda Brand 860365C

Synonym(s):

1,2-distearoyl-d70-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C44H18NO8PD70
CAS Number:
Molecular Weight:
860.58
UNSPSC Code:
12352100
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (860365C-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860365C

concentration

10 mg/mL (860365C-10mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H]

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1/i1D3,2D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2,32D2,33D2,34D2,35D2,36D2,37D2

InChI key

NRJAVPSFFCBXDT-MNVUKWGGSA-N

General description

18:0 Phosphocholine (PC)-deuterium70 contains phosphocholine head group and an 18-deuterated carbon chain.[1]
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.
Phosphatidylcholine (PC) is the most common phospholipid in mammalian membranes.[2] It is a strong bilayer-forming lipid,[2] which is a crucial component of the mucosal layer of the colon.[3]

Biochem/physiol Actions

Phosphatidylcholine (PC) acts as a surfactant within the mucus to form a hydrophobic surface, which prevents bacterial invasion.[3] It reduces the levels of cholesterol and triglycerides. Phosphatidylcholine is used to treat fat embolism.[4]
Phosphocholine (PC) post-translationally alter the placental neurokinin B hormone and decrease the host immune response.[5]

Packaging

5 mL Clear Glass Sealed Ampule (860365C-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book (2017)
The Membranes of Cells (2016)
Identification of a novel mammalian post-translational modification, phosphocholine, on placental secretory polypeptides
Lovell TM, et al.
Journal of Molecular Endocrinology, 39 (2007)
Estimation of bacterial biomass in subsurface sediments by quantifying intact membrane phospholipids
Zink K, et al.
Analytical and Bioanalytical Chemistry, 390(3), 885-896 (2008)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service