Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

860203C

Avanti

18:1 Phosphatidylbutanol

1,2-dioleoyl-sn-glycero-3-phosphobutanol (sodium salt), chloroform

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-phosphobutanol (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C43H80O8PNa
CAS Number:
Molecular Weight:
779.05
UNSPSC Code:
12352211
NACRES:
NA.25

assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (860203C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860203C

concentration

10 mg/mL (860203C-25mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C43H81O8P.Na/c1-4-7-10-12-14-16-18-20-22-24-26-28-30-32-34-36-42(44)48-39-41(40-50-52(46,47)49-38-9-6-3)51-43(45)37-35-33-31-29-27-25-23-21-19-17-15-13-11-8-5-2;/h20-23,41H,4-19,24-40H2,1-3H3,(H,46,47);/q;+1/p-1/b22-20-,23-21-;/t41-;/m1./s1

InChI key

BXQFBLMPDPPGFW-RLWYHRQISA-M

General description

Phosphatidylbutanol is a low polarity phosphatidylalcohol. This phospholipid is synthesized by the phospholipase D (PLD) catalyzed transphosphatidylation reaction of phosphatidylcholine (PC) and butanol.[1]

Application

18:1 Phosphatidylbutanol or 1,2-dioleoyl-sn-glycero-3-phosphobutanol (sodium salt) has been used as a standard to measure phospholipase D (PLD) activity in smooth muscle cells (SMCs) and cancer cell lines by thin layer-chromatography.[2][3][4]

Biochem/physiol Actions

Phosphatidylbutanol is used in phospholipase D activity assays as a potential biomarker.[1]

Packaging

5 mL Clear Glass Sealed Ampule (860203C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elyssa Bernfeld et al.
The Journal of biological chemistry, 293(42), 16390-16401 (2018-09-09)
Glutamine is a key nutrient required for sustaining cell proliferation, contributing to nucleotide, protein, and lipid synthesis. The mTOR complex 1 (mTORC1) is a highly conserved protein complex that acts as a sensor of nutrients, relaying signals for the shift
Y Liu et al.
American journal of physiology. Lung cellular and molecular physiology, 295(3), L471-L478 (2008-07-16)
We have previously reported the participation of mitogen-activated protein, Rho, and phosphoinositide-3 (PI3) kinases in separate pathways in serotonin (5-HT)-induced proliferation of pulmonary artery smooth muscle cells (SMCs). In this study, we investigated the possible participation of phospholipase D (PLD)
Mikhail Vyssotski et al.
Lipids, 44(4), 381-389 (2008-12-17)
High-performance TLC and (31)P-NMR were assessed as methods of observing the presence of numerous low polarity phospholipids: bis-phosphatidic acid (BPA), semi-lyso bis-phosphatidic acid (SLBPA), N-acyl phosphatidylethanolamine (NAPE), N-(1,1-dimethyl-3-oxo-butyl)-phosphatidylethanolamine (diacetone adduct of PE, DOBPE), N-acetyl PE, phosphatidylmethanol (PM), phosphatidylethanol (PEt), phosphatidyl-n-propanol
Maria A Frias et al.
The Journal of biological chemistry, 295(1), 263-274 (2019-11-27)
Mammalian target of rapamycin complex 1 (mTORC1) promotes cell growth and proliferation in response to nutrients and growth factors. Amino acids induce lysosomal translocation of mTORC1 via the Rag GTPases. Growth factors activate Ras homolog enriched in brain (Rheb), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service