Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

857520P

Avanti

N-Hexanoyl-L-homoserine lactone

Avanti Research - A Croda Brand

Synonym(s):

(S)-N-(2-Oxotetrahydrofuran-3-yl)hexanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
UNSPSC Code:
12352211
NACRES:
NA.25
Product 857520P is not currently sold in your country. Contact Technical Service

description

(S)-N-(2-Oxotetrahydrofuran-3-yl)hexanamide

assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (857520P-10MG)
pkg of 1 × 50 mg (857520P-50MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C1OCC[C@@H]1NC(CCCCC)=O

InChI

1S/C10H17NO3/c1-2-3-4-5-9(12)11-8-6-7-14-10(8)13/h8H,2-7H2,1H3,(H,11,12)/t8-/m0/s1

InChI key

ZJFKKPDLNLCPNP-QMMMGPOBSA-N

General description

N-Hexanoyl-L-homoserine lactone is produced and utilized by various Gram-negative bacteria as a quorum sensing (QS) signal molecule.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (857520P-10MG and 857520P-50MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurie Delalande et al.
FEMS microbiology ecology, 52(1), 13-20 (2005-12-07)
The half-life of N-hexanoyl-l-homoserine lactone (C6-HSL) was determined under various pH and temperature conditions, and in several plant environments. C6-HSL was sensitive to alkaline pH, a process that was also temperature-dependent. In addition, C6-HSL disappeared from plant environments, i.e. axenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service